siRNA Details
Gene Symbol: ZNF534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143938.1 | NP_001137410.1 | 4 | 683 | ||
NM_001143939.1 | NP_001137411.1 | 4 | 722 | |||
GenBank mRNA | AB086839.1 | - | 4 | 683 | ||
AB086840.1 | - | 4 | 722 |
