siRNA Details
Gene Symbol: SMIM17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193628.1 | NP_001180557.1 | 2 | 272 | ||
XM_017026303.1 | XP_016881792.1 | 3 | 1295 | |||
GenBank mRNA | AK095199.1 | - | 2 | 278 |
