siRNA Details
Gene Symbol: METTL11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136107.1 | NP_001129579.1 | 3 | 577 | ||
XM_011509232.2 | XP_011507534.1 | 4 | 598 | |||
XM_011509233.2 | XP_011507535.1 | 5 | 564 | |||
XM_011509234.2 | XP_011507536.1 | 4 | 576 | |||
GenBank mRNA | BC157860.1 | - | 3 | 577 | 9057079 | |
BC171858.1 | - | 3 | 577 | 9054512 |
