siRNA Details
Gene Symbol: LOC150051 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321023.1 | NP_001307952.1 | 1 | 255 | ||
GenBank mRNA | BC014150.1 | - | 1 | 255 | ||
CA430855.1 | - | 2 | 580 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC150051 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321023.1 | NP_001307952.1 | 1 | 255 | ||
GenBank mRNA | BC014150.1 | - | 1 | 255 | ||
CA430855.1 | - | 2 | 580 |