siRNA Details
Gene Symbol: LINC01107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037809.1 | 3 | 1947 | |||
GenBank mRNA | AK057376.1 | - | 3 | 1503 | ||
BC101453.1 | - | 1 | 264 | |||
BC101454.1 | - | 1 | 264 | |||
BC101455.1 | - | 1 | 264 | |||
BC101456.1 | - | 1 | 264 |
