siRNA Details
Gene Symbol: LOC151174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026925.1 | 2 | 390 | |||
NR_026926.1 | 2 | 390 | ||||
GenBank mRNA | AK057399.1 | - | 2 | 326 | ||
CD367561.1 | - | 2 | 464 | |||
DA518680.1 | - | 2 | 390 |
