siRNA Details
Gene Symbol: LOC151174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026925.1 | 3 | 1579 | |||
NR_026926.1 | 2, 3 | 466 | ||||
GenBank mRNA | AF451986.1 | - | 1, 2 | 57 | ||
AK057399.1 | - | 3 | 1513 | |||
BU739907.1 | - | 1, 2 | 395 |
