siRNA Details
Gene Symbol: LOC151174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026925.1 | 3 | 1802 | |||
NR_026926.1 | 3 | 689 | ||||
GenBank mRNA | AF451986.1 | - | 2 | 280 | ||
AK057399.1 | - | 3 | 1736 | |||
BU739907.1 | - | 1 | 172 | |||
CD367561.1 | - | 1 | 165 |
