siRNA Details
Gene Symbol: LOC100289673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033175.1 | 3 | 526 | |||
GenBank mRNA | AI125295.1 | - | 1 | 123 | ||
AI216275.1 | - | 1 | 141 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100289673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033175.1 | 3 | 526 | |||
GenBank mRNA | AI125295.1 | - | 1 | 123 | ||
AI216275.1 | - | 1 | 141 |