siRNA Details
Gene Symbol: CXorf51B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244892.1 | NP_001231821.1 | 1, 2 | 151 |
Gene Symbol: CXorf51A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144064.2 | NP_001137536.1 | 1, 2 | 151 | ||
GenBank mRNA | AA417208.1 | - | 1, 2 | 151 |
