siRNA Details
Gene Symbol: ZNF846 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077624.1 | NP_001071092.1 | 6 | 1841 | ||
XM_005259772.3 | XP_005259829.1 | 6 | 1632 | |||
XM_005259773.4 | XP_005259830.1 | 6 | 1838 | |||
XM_005259775.3 | XP_005259832.1 | 6 | 1629 | |||
XM_011527717.2 | XP_011526019.1 | 6 | 1644 | |||
XM_011527718.2 | XP_011526020.1 | 6 | 1641 | |||
XM_011527719.2 | XP_011526021.1 | 6 | 1660 | |||
XM_011527720.2 | XP_011526022.1 | 5 | 1415 | |||
XM_011527721.2 | XP_011526023.1 | 4 | 1189 | |||
XM_017026404.1 | XP_016881893.1 | 6 | 1799 | |||
XM_017026405.1 | XP_016881894.1 | 5 | 2281 | |||
XM_017026406.1 | XP_016881895.1 | 3 | 1119 | |||
GenBank mRNA | BC118576.1 | - | 6 | 1412 | 40053684 | |
BC118657.1 | - | 5 | 1329 |
