siRNA Details
Gene Symbol: ZNF846 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077624.1 | NP_001071092.1 | 5, 6 | 831 | ||
XM_005259772.3 | XP_005259829.1 | 5, 6 | 622 | |||
XM_005259773.4 | XP_005259830.1 | 5, 6 | 828 | |||
XM_005259775.3 | XP_005259832.1 | 5, 6 | 619 | |||
XM_006722658.3 | XP_006722721.1 | 5, 6 | 831 | |||
XM_011527717.2 | XP_011526019.1 | 5, 6 | 634 | |||
XM_011527718.2 | XP_011526020.1 | 5, 6 | 631 | |||
XM_011527719.2 | XP_011526021.1 | 5, 6 | 650 | |||
XM_011527720.2 | XP_011526022.1 | 4, 5 | 405 | |||
XM_011527721.2 | XP_011526023.1 | 3, 4 | 179 | |||
XM_017026404.1 | XP_016881893.1 | 5, 6 | 789 | |||
XM_017026407.1 | XP_016881896.1 | 5, 6 | 831 | |||
XM_017026408.1 | XP_016881897.1 | 5, 6 | 522 | |||
GenBank mRNA | BC118576.1 | - | 5, 6 | 402 | 40053684 |
