siRNA Details
Gene Symbol: LRRC73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012974.2 | NP_001012992.1 | 3 | 1092 | ||
NM_001271882.1 | NP_001258811.1 | 3 | 400 | |||
GenBank mRNA | BC107130.2 | - | 3 | 300 | ||
BC108661.1 | - | 3 | 1092 | 5240367 | ||
HY024930.1 | - | 3 | 400 |
