siRNA Details
Gene Symbol: ZSCAN23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012455.1 | NP_001012458.1 | 4 | 1190 | ||
XM_005248950.2 | XP_005249007.1 | 4 | 1188 | |||
XM_011514406.2 | XP_011512708.1 | 4 | 1188 | |||
XM_011514408.2 | XP_011512710.1 | 5 | 1375 | |||
XM_011514410.2 | XP_011512712.1 | 4 | 1188 | |||
XM_017010479.1 | XP_016865968.1 | 4 | 4404 | |||
GenBank mRNA | AK092117.1 | - | 3 | 1020 | ||
AK309235.1 | - | 2 | 507 | |||
AL833347.1 | - | 4 | 1155 | |||
BC101557.1 | - | 3 | 1092 | 8069063 | ||
BC101559.1 | - | 3 | 1092 | 8069065 |
