siRNA Details
Gene Symbol: TTTY7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001534.2 | 4 | 671 | |||
GenBank mRNA | AF278762.1 | - | 1 | 170 |
Gene Symbol: TTTY7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003592.1 | 4 | 671 | |||
GenBank mRNA | BX089948.1 | - | 1 | 169 |
