siRNA Details
Gene Symbol: LINC01018 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024423.2 | 3 | 407 | |||
NR_024424.2 | 1 | 421 | ||||
GenBank mRNA | AK125283.1 | - | 3 | 369 | ||
BC043253.2 | - | 1 | 105 | |||
BC106918.2 | - | 1 | 400 | |||
BC106919.1 | - | 1 | 403 | |||
BI602592.1 | - | 1 | 425 | |||
DA240476.1 | - | 2 | 560 |
