siRNA Details
Gene Symbol: LMNTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173573.2 | NP_775844.2 | 8 | 945 | ||
XM_011519965.2 | XP_011518267.2 | 8 | 1120 | |||
XM_011519967.2 | XP_011518269.2 | 8 | 1120 | |||
XM_017017476.1 | XP_016872965.1 | 8 | 1120 | |||
XM_017017477.1 | XP_016872966.1 | 8 | 1105 | |||
XM_017017478.1 | XP_016872967.1 | 8 | 961 | |||
XM_017017479.1 | XP_016872968.1 | 8 | 1120 | |||
GenBank mRNA | BC039077.1 | - | 8 | 945 | 5168905 |
