siRNA Details
Gene Symbol: DSCR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026719.2 | 3 | 1312 | |||
GenBank mRNA | AB066100.1 | - | 1 | 1079 | ||
AK313458.1 | - | 1 | 817 | |||
BC029827.1 | - | 1 | 650 | |||
BC066653.1 | - | 3 | 1312 |
