siRNA Details
Gene Symbol: LINC01366 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026945.2 | 3 | 706 | |||
NR_108024.1 | 3 | 687 | ||||
NR_108025.1 | 2 | 616 | ||||
GenBank mRNA | AK093725.1 | - | 3 | 1402 | ||
BC093717.1 | - | 1 | 179 | |||
BC111994.1 | - | 1 | 179 | |||
BQ053612.1 | - | 3 | 668 | |||
CD518314.1 | - | 3 | 651 |
