siRNA Details
Gene Symbol: FLJ37201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026835.1 | 2 | 1276 | |||
GenBank mRNA | AK094520.1 | - | 1 | 431 | ||
BC043217.2 | - | 2 | 1276 | |||
BC104181.2 | - | 1 | 430 | |||
BC104182.2 | - | 1 | 430 | |||
BC112983.1 | - | 1 | 430 | |||
BC112984.1 | - | 1 | 430 |
