siRNA Details
Gene Symbol: KIRREL3-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040078.1 | 1 | 731 | |||
GenBank mRNA | AK097543.1 | - | 1 | 731 | ||
BC113593.1 | - | 1 | 238 | 8322645 | ||
BC113597.1 | - | 1 | 238 | 8322649 |
