siRNA Details
Gene Symbol: LOC283278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017018657.1 | XP_016874146.1 | 1 | 168 | ||
XM_017018658.1 | XP_016874147.1 | 2 | 156 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC283278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017018657.1 | XP_016874146.1 | 1 | 168 | ||
XM_017018658.1 | XP_016874147.1 | 2 | 156 |