siRNA Details
Gene Symbol: LINC01198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120426.1 | 4, 5 | 563 | |||
GenBank mRNA | AW269438.1 | - | 2 | 370 |
Gene Symbol: LOC102723968 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130763.1 | 4, 5 | 1156 | |||
GenBank mRNA | AK098560.1 | - | 4, 5 | 1156 | ||
AK127969.1 | - | 3, 4 | 2170 |
