siRNA Details
Gene Symbol: TMEM235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204210.1 | NP_001191139.1 | 5 | 1199 | ||
NM_001204211.1 | NP_001191140.1 | 4 | 1118 | |||
NM_001204212.1 | NP_001191141.1 | 3 | 980 | |||
XM_011524620.2 | XP_011522922.1 | 6 | 737 | |||
XM_011524624.2 | XP_011522926.1 | 4 | 575 | |||
XM_017024459.1 | XP_016879948.1 | 5 | 650 | |||
XM_017024460.1 | XP_016879949.1 | 6 | 737 | |||
GenBank mRNA | BC042066.1 | - | 3 | 980 | ||
BC148207.2 | - | 3 | 807 | |||
BI757060.1 | - | 4 | 537 | |||
BI757981.1 | - | 4 | 469 |
