siRNA Details
Gene Symbol: LINC00670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034144.1 | 2 | 269 | |||
NR_034145.1 | 3 | 431 | ||||
GenBank mRNA | AK092009.1 | - | 3 | 430 | ||
BC122562.1 | - | 2 | 256 | |||
BC122870.1 | - | 2 | 256 | |||
DB255288.1 | - | 3 | 432 |
