siRNA Details
Gene Symbol: FAM27E5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028336.1 | 2 | 377 | |||
GenBank mRNA | BC031617.1 | - | 2 | 377 | 5169482 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM27E5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028336.1 | 2 | 377 | |||
GenBank mRNA | BC031617.1 | - | 2 | 377 | 5169482 |