siRNA Details
Gene Symbol: PHLDB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198850.3 | NP_942147.3 | 3 | 484 | ||
XM_005259169.3 | XP_005259226.1 | 3 | 567 | |||
XM_005259170.3 | XP_005259227.1 | 3 | 567 | |||
XM_005259171.3 | XP_005259228.1 | 3 | 567 | |||
XM_011527236.2 | XP_011525538.1 | 3 | 671 | |||
GenBank mRNA | AK097512.1 | - | 3 | 294 | ||
BC029299.1 | - | 3 | 484 | |||
BC070094.1 | - | 3 | 586 | 4374205 |
