siRNA Details
Gene Symbol: MIR7-3HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027148.1 | 2 | 290 | |||
GenBank mRNA | AB058892.2 | - | 2 | 255 | ||
AB058893.2 | - | 2 | 255 | |||
BC090884.1 | - | 2 | 254 | |||
BC117294.1 | - | 2 | 228 | |||
BC117296.1 | - | 2 | 228 |
