siRNA Details
Gene Symbol: ZNF493 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076678.2 | NP_001070146.1 | 4 | 2053 | ||
NM_175910.6 | NP_787106.4 | 2 | 1830 | |||
GenBank mRNA | AB209086.1 | - | 2 | 1798 | ||
AK093823.1 | - | 1 | 423 | |||
AK095616.1 | - | 1 | 582 | |||
AK127476.1 | - | 1 | 1438 | |||
AK128494.1 | - | 2 | 1816 | |||
BC112346.1 | - | 1 | 1649 | |||
BX648842.1 | - | 4 | 2043 |
