siRNA Details
Gene Symbol: SMG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005245648.3 | XP_005245705.1 | 1 | 14 | ||
XM_005245649.3 | XP_005245706.1 | 1 | 14 | |||
XM_006711676.3 | XP_006711739.1 | 1 | 14 | |||
XM_011510205.2 | XP_011508507.1 | 1 | 14 | |||
XM_011510206.2 | XP_011508508.1 | 1 | 14 | |||
XM_011510207.2 | XP_011508509.1 | 1 | 14 | |||
XM_011510208.2 | XP_011508510.1 | 1 | 14 | |||
XM_011510209.2 | XP_011508511.1 | 1 | 14 |
Gene Symbol: SMG7-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL050020.1 | - | 1 | 291 |
