siRNA Details
Gene Symbol: LINC00486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027098.1 | 5 | 616 | |||
NR_027100.1 | 4 | 353 | ||||
GenBank mRNA | AK095182.1 | - | 5 | 616 | ||
BC121144.1 | - | 4 | 353 | |||
BC121145.1 | - | 4 | 356 |
