siRNA Details
Gene Symbol: LINC01096 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015450.1 | 1 | 716 | |||
GenBank mRNA | AK058104.1 | - | 1, 2 | 230 | ||
BC035174.2 | - | 1 | 685 | |||
BC048110.1 | - | 1 | 693 | |||
BC053955.1 | - | 1 | 693 |
