siRNA Details
Gene Symbol: LOC285638 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL833206.1 | - | 4 | 4933 |
Gene Symbol: THEMIS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011535813.2 | XP_011534115.1 | 8 | 5184 |
