siRNA Details
Gene Symbol: CFAP157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012502.2 | NP_001012520.2 | 7 | 1279 | ||
XM_006717064.3 | XP_006717127.2 | 7 | 2007 | |||
XM_011518559.2 | XP_011516861.2 | 7 | 2010 | |||
XM_017014630.1 | XP_016870119.1 | 7 | 1346 | |||
GenBank mRNA | AK094948.1 | - | 6 | 1542 | ||
AK126437.1 | - | 3 | 1315 | |||
AL833241.1 | - | 1 | 474 | |||
BC133027.1 | - | 1 | 921 | |||
BC141809.1 | - | 7 | 1356 | 40147564 |
