siRNA Details
Gene Symbol: LOC338797 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103736.1 | 3 | 617 | |||
GenBank mRNA | AK098523.1 | - | 3 | 617 | ||
BC093897.1 | - | 1 | 145 | 7939742 | ||
BC093899.1 | - | 1 | 145 | 7939744 |
