siRNA Details
Gene Symbol: THEG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205273.2 | NP_001192202.1 | 2 | 286 | ||
NM_001278575.1 | NP_001265504.1 | 3 | 696 | |||
NM_001278576.1 | NP_001265505.1 | 4 | 820 | |||
NM_001278577.1 | NP_001265506.1 | 4 | 831 | |||
NM_001278578.1 | NP_001265507.1 | 3 | 689 | |||
XM_011526302.2 | XP_011524604.1 | 3 | 2475 | |||
XM_011526303.2 | XP_011524605.1 | 3 | 1300 | |||
XM_017026126.1 | XP_016881615.1 | 3 | 2811 | |||
XM_017026127.1 | XP_016881616.1 | 3 | 2270 | |||
XM_017026128.1 | XP_016881617.1 | 3 | 2468 | |||
XM_017026129.1 | XP_016881618.1 | 3 | 1307 | |||
XM_017026130.1 | XP_016881619.1 | 3 | 1032 | |||
XM_017026131.1 | XP_016881620.1 | 4 | 638 | |||
XM_017026132.1 | XP_016881621.1 | 3 | 831 | |||
XM_017026133.1 | XP_016881622.1 | 2 | 499 | |||
GenBank mRNA | BC043286.1 | - | 4 | 759 | ||
FJ211597.1 | - | 2 | 286 |
Gene Symbol: LOC101927277 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR627061.1 | - | 2 | 835 |
