siRNA Details
Gene Symbol: CLEC4G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244856.1 | NP_001231785.1 | 8, 9 | 780 | ||
NM_198492.3 | NP_940894.1 | 8, 9 | 816 | |||
GenBank mRNA | AY203939.1 | - | 8, 9 | 737 | ||
AY358431.1 | - | 9, 10 | 813 | |||
AY423706.1 | - | 8, 9 | 816 | |||
BC039679.1 | - | 4, 5 | 403 | |||
BC093691.1 | - | 8, 9 | 770 | 7939536 | ||
BC093693.1 | - | 8, 9 | 770 | 7939538 | ||
BC143309.1 | - | 8, 9 | 734 | 9051814 |
