siRNA Details
Gene Symbol: LINC01140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026985.1 | 1 | 215 | |||
NR_026986.1 | 1 | 215 | ||||
NR_026987.1 | 1 | 215 | ||||
GenBank mRNA | AL833352.1 | - | 1 | 138 | ||
BC041867.1 | - | 1 | 186 | |||
BC126240.1 | - | 1 | 144 | 8991956 | ||
BC132861.1 | - | 1 | 144 | |||
DA398164.1 | - | 1 | 202 | |||
DB100426.1 | - | 1 | 184 | |||
DB462107.2 | - | 1 | 218 |
