siRNA Details
Gene Symbol: PRAMEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294139.1 | NP_001281068.1 | 1, 2 | 276 | ||
GenBank mRNA | BC040902.1 | - | 1, 2 | 276 |
Gene Symbol: PRAMEF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011541458.1 | XP_011539760.1 | 1, 2 | 290 |
Gene Symbol: PRAMEF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542078.2 | XP_011540380.1 | 1, 2 | 291 |
Gene Symbol: PRAMEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542004.1 | XP_011540306.1 | 1, 2 | 291 |
