siRNA Details
Gene Symbol: C2orf70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105519.2 | NP_001098989.1 | 2, 3 | 408 | ||
NM_001322426.1 | NP_001309355.1 | 3, 4 | 444 | |||
XM_011532810.2 | XP_011531112.1 | 4, 5 | 611 | |||
XM_011532811.2 | XP_011531113.1 | 4, 5 | 2107 | |||
XM_017003962.1 | XP_016859451.1 | 4, 5 | 462 | |||
XM_017003963.1 | XP_016859452.1 | 2, 3 | 406 | |||
XM_017003964.1 | XP_016859453.1 | 3, 4 | 438 | |||
XM_017003965.1 | XP_016859454.1 | 4, 5 | 462 | |||
XM_017003966.1 | XP_016859455.1 | 4, 5 | 611 | |||
GenBank mRNA | AA725022.1 | - | 2, 3 | 343 | ||
AK091483.1 | - | 2, 3 | 355 | |||
BX114846.1 | - | 3, 4 | 417 | |||
HY021315.1 | - | 3, 4 | 447 |
