siRNA Details
Gene Symbol: LINC01021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038848.1 | 2, 3 | 397 | |||
GenBank mRNA | BC046201.1 | - | 2, 3 | 397 | ||
BC101982.1 | - | 2, 3 | 382 | |||
CQ819101.1 | - | 2, 3 | 405 |
