siRNA Details
Gene Symbol: LINC01021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038848.1 | 3 | 511 | |||
GenBank mRNA | BC046201.1 | - | 3 | 511 | ||
BC101982.1 | - | 3 | 496 | |||
CQ819099.1 | - | 4 | 591 | |||
CQ819101.1 | - | 3 | 519 |
