siRNA Details
Gene Symbol: C16orf96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145011.1 | NP_001138483.1 | 10 | 2662 | ||
XM_005255298.3 | XP_005255355.1 | 11 | 3189 | |||
XM_005255301.3 | XP_005255358.1 | 9 | 3084 | |||
XM_011522475.2 | XP_011520777.1 | 12 | 3172 |
