siRNA Details
Gene Symbol: ANKRD62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277333.1 | NP_001264262.1 | 10 | 1346 | ||
XM_017025720.1 | XP_016881209.1 | 10 | 1399 | |||
GenBank mRNA | BX649021.1 | - | 5 | 441 | ||
DW010010.1 | - | 6 | 463 | |||
DW010011.1 | - | 4 | 626 |
