siRNA Details
Gene Symbol: ZNF850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193552.1 | NP_001180481.1 | 4, 5 | 392 | ||
GenBank mRNA | BC052603.1 | - | 4, 5 | 392 | ||
BC136266.1 | - | 4, 5 | 330 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193552.1 | NP_001180481.1 | 4, 5 | 392 | ||
GenBank mRNA | BC052603.1 | - | 4, 5 | 392 | ||
BC136266.1 | - | 4, 5 | 330 |