siRNA Details
Gene Symbol: LOC105371448 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003084.1 | XP_016858573.1 | 5 | 2370 | ||
XM_017003085.1 | XP_016858574.1 | 4 | 2456 |
Gene Symbol: LOC343052 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CN286844.1 | - | 2 | 194 |
