siRNA Details
Gene Symbol: ANKRD20A12P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046228.1 | 5 | 1318 | |||
GenBank mRNA | AK090412.1 | - | 5 | 2319 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ANKRD20A12P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046228.1 | 5 | 1318 | |||
GenBank mRNA | AK090412.1 | - | 5 | 2319 |