siRNA Details
Gene Symbol: PCDH15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142771.1 | NP_001136243.1 | 35 | 5076 | ||
NM_001142772.1 | NP_001136244.1 | 34 | 5061 | |||
XM_017016571.1 | XP_016872060.1 | 37 | 5782 | |||
XM_017016572.1 | XP_016872061.1 | 34 | 5651 | |||
GenBank mRNA | EU718482.1 | - | 34 | 5055 |
