siRNA Details
Gene Symbol: TMEM233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136534.1 | NP_001130006.1 | 2 | 677 | ||
XM_005253880.3 | XP_005253937.1 | 2 | 2013 | |||
XM_005253881.3 | XP_005253938.1 | 2 | 2009 | |||
XM_011538328.2 | XP_011536630.1 | 2 | 2010 | |||
XM_011538329.2 | XP_011536631.1 | 2 | 2010 | |||
XM_011538330.2 | XP_011536632.1 | 2 | 2010 | |||
XM_011538331.2 | XP_011536633.1 | 2 | 2008 | |||
GenBank mRNA | AK296289.1 | - | 2 | 602 |
