siRNA Details
Gene Symbol: MCF2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011537482.1 | XP_011535784.1 | 1, 2 | 65 | ||
XM_011537483.2 | XP_011535785.1 | 1, 2 | 185 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MCF2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011537482.1 | XP_011535784.1 | 1, 2 | 65 | ||
XM_011537483.2 | XP_011535785.1 | 1, 2 | 185 |